Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat Submandibular Glandular Kallikrein 9 ELISA Kit

Supplier: Antibodies.com LLC
A80057-96
76730-074EA 605 USD InStock
76730-074
Rat Submandibular Glandular Kallikrein 9 ELISA Kit
Assays ELISAs
Rat Submandibular Glandular Kallikrein 9 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Submandibular Glandular Kallikrein 9 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 31.25 - 2000 pg/ml
  • Sensitivity: 18.75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat Submandibular Glandular Kallikrein 9 ELISA kit (A80057) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Submandibular Glandular Kallikrein 9 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Submandibular Glandular Kallikrein 9 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Submandibular Glandular Kallikrein 9 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Submandibular Glandular Kallikrein 9 Antibody, which binds the captured Submandibular Glandular Kallikrein 9 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Submandibular Glandular Kallikrein 9 captured in each well. The concentration of Submandibular Glandular Kallikrein 9 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty