Would you like to log in again?
Human IgM ELISA Matched Antibody Pair Kit
Supplier: Antibodies.com LLC
A290886-1
76730-146EA 1205 USD InStock
76730-146
Human IgM ELISA Matched Antibody Pair Kit
Assays ELISAs
This kit can be used for the detection of therapeutic antibodies, from samples obtained from experimental animals, to provide accurate pharmacokinetic data.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty