Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Transferrin Receptor 2/TFR2 ELISA Kit

Supplier: Antibodies.com LLC
A78866-96
76730-924EA 605 USD InStock
76730-924
Human Transferrin Receptor 2/TFR2 ELISA Kit
Assays ELISAs
Human Transferrin receptor 2/TFR2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Transferrin receptor 2/TFR2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Transferrin Receptor 2 / TFR2 ELISA Kit (A78866) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Transferrin Receptor 2 / TFR2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Transferrin Receptor 2 / TFR2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Transferrin Receptor 2 / TFR2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Transferrin Receptor 2 / TFR2 Antibody, which binds the captured Transferrin Receptor 2 / TFR2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Transferrin Receptor 2 / TFR2 captured in each well. The concentration of Transferrin Receptor 2 / TFR2 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty