Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat Secretin/SCT ELISA Kit

Supplier: Antibodies.com LLC
A80363-96
76730-928EA 605 USD InStock
76730-928
Rat Secretin/SCT ELISA Kit
Assays ELISAs
Rat Secretin/SCT ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Secretin/SCT in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Secretin/SCT ELISA kit (A80363) employs the competitive enzyme immunoassay technique for the quantitative measurement of rat Secretin/SCT in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Secretin/SCT antigen. During the incubation, Secretin/SCT present in the samples or standards competes with the fixed amount of immobilized Secretin/SCT for binding sites on the Biotinylated Anti-Secretin/SCT Antibody. The more Secretin/SCT present in a sample or standard, the less Biotinylated Anti-Secretin/SCT Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Secretin/SCT Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Secretin/SCT present in each sample or standard. The concentration of Secretin/SCT can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty