Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Porcine Endothelin 1 ELISA Kit

Supplier: Antibodies.com LLC
A74780-96
76731-972EA 705 USD InStock
76731-972
Porcine Endothelin 1 ELISA Kit
Assays ELISAs
Porcine Endothelin 1 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of porcine Endothelin 1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 1.25 - 80 pg/ml
  • Sensitivity: 0.625 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Porcine Endothelin 1 ELISA kit (A74780) employs the competitive enzyme immunoassay technique for the quantitative measurement of porcine Endothelin 1 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Endothelin 1 antigen. During the incubation, Endothelin 1 present in the samples or standards competes with the fixed amount of immobilized Endothelin 1 for binding sites on the Biotinylated Anti-Endothelin 1 Antibody. The more Endothelin 1 present in a sample or standard, the less Biotinylated Anti-Endothelin 1 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Endothelin 1 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Endothelin 1 present in each sample or standard. The concentration of Endothelin 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty