Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat Peptide YY/PYY ELISA Kit

Supplier: Antibodies.com LLC
A80361-96
76731-992EA 605 USD InStock
76731-992
Rat Peptide YY/PYY ELISA Kit
Assays ELISAs
Rat Peptide YY/PYY ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Peptide YY/PYY in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 15.625 - 1000 pg/ml
  • Sensitivity: 9.375 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat Peptide YY/PYY ELISA kit (A80361) employs the competitive enzyme immunoassay technique for the quantitative measurement of rat Peptide YY/PYY in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Peptide YY/PYY antigen. During the incubation, Peptide YY/PYY present in the samples or standards competes with the fixed amount of immobilized Peptide YY/PYY for binding sites on the Biotinylated Anti-Peptide YY/PYY Antibody. The more Peptide YY/PYY present in a sample or standard, the less Biotinylated Anti-Peptide YY/PYY Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Peptide YY/PYY Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Peptide YY/PYY present in each sample or standard. The concentration of Peptide YY/PYY can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty