Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cGMP ELISA Kit

Supplier: Antibodies.com LLC
A303893-96
76737-172EA 745 USD InStock
76737-172
cGMP ELISA Kit
Assays ELISAs
cGMP ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of cGMP in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 0.781 - 50 pmol/ml
  • Sensitivity: 0.469 pmol/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

cGMP ELISA kit (A303893) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal cGMP in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with cGMP antigen. During the incubation, cGMP present in the samples or standards competes with the fixed amount of immobilized cGMP for binding sites on the Biotinylated anti-cGMP antibody. The more cGMP present in a sample or standard, the less Biotinylated anti-cGMP antibody that binds to the plate. Following incubation, unbound Biotinylated anti-cGMP antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of cGMP present in each sample or standard. The concentration of cGMP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty