Would you like to log in again?
Sheep Neurokinin-B ELISA Kit
Supplier: Antibodies.com LLC
A303865-96
76737-898EA 935 USD InStock
76737-898
Sheep Neurokinin-B ELISA Kit
Assays ELISAs
Sheep Neurokinin-B ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of sheep Neurokinin-B in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Sheep Neurokinin-B ELISA kit (A303865) employs the competitive enzyme immunoassay technique for the quantitative measurement of sheep Neurokinin-B in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Neurokinin-B antigen. During the incubation, Neurokinin-B present in the samples or standards competes with the fixed amount of immobilized Neurokinin-B for binding sites on the Biotinylated Anti-Neurokinin-B Antibody. The more Neurokinin-B present in a sample or standard, the less Biotinylated Anti-Neurokinin-B Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Neurokinin-B Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Neurokinin-B present in each sample or standard. The concentration of Neurokinin-B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty