Would you like to log in again?
Human 5-HETE ELISA Kit
Supplier: Antibodies.com LLC
A74628-96
76737-902EA 605 USD InStock
76737-902
Human 5-HETE ELISA Kit
Assays ELISAs
This Human 5-HETE ELISA kit is an in vitro competitive enzyme-linked immunosorbent assay for the quantitative measurement of 5-HETE in plasma, tissue homogenates and other biological fluids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty