Would you like to log in again?
Rat Klk8 ELISA Kit
Supplier: Antibodies.com LLC
A80056-96
76738-836EA 605 USD InStock
76738-836
Rat Klk8 ELISA Kit
Assays ELISAs
This Rat Klk8 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat Klk8 in plasma, tissue homogenates and other biological fluids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty