Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sheep Antidiuretic Hormone ELISA Kit

Supplier: Antibodies.com LLC
A87523-96
76738-990EA 705 USD InStock
76738-990
Sheep Antidiuretic Hormone ELISA Kit
Assays ELISAs
Sheep Antidiuretic Hormone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of sheep Antidiuretic Hormone in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 15.625 to 1000 pg/ml
  • Sensitivity: 9.375 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Sheep Antidiuretic Hormone ELISA kit (A87523) employs the competitive enzyme immunoassay technique for the quantitative measurement of sheep Antidiuretic Hormone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Antidiuretic Hormone antigen. During the incubation, Antidiuretic Hormone present in the samples or standards competes with the fixed amount of immobilized Antidiuretic Hormone for binding sites on the Biotinylated Anti-Antidiuretic Hormone Antibody. The more Antidiuretic Hormone present in a sample or standard, the less Biotinylated Anti-Antidiuretic Hormone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Antidiuretic Hormone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Antidiuretic Hormone present in each sample or standard. The concentration of Antidiuretic Hormone can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty