Would you like to log in again?
Human Herpes Simplex Virus 2 IgG ELISA Kit
Supplier: Antibodies.com LLC
A303135-96
76739-378EA 745 USD InStock
76739-378
Human Herpes Simplex Virus 2 IgG ELISA Kit
Assays ELISAs
Human Herpes Simplex Virus 2 IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Herpes Simplex Virus 2 IgG in serum, plasma, tissue homogenates, and other biological fluids.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty