Would you like to log in again?
Human PI3 Kinase p110 Beta ELISA Kit
Supplier: Antibodies.com LLC
A247135-96
76741-422EA 605 USD InStock
76741-422
Human PI3 Kinase p110 Beta ELISA Kit
Assays ELISAs
Human PI3 Kinase p110 beta ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PI3 Kinase p110 beta in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human PI3 Kinase p110 beta ELISA kit (A247135) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PI3 Kinase p110 beta in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PI3 Kinase p110 beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PI3 Kinase p110 beta present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PI3 Kinase p110 beta Antibody, which binds the captured PI3 Kinase p110 beta present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PI3 Kinase p110 beta captured in each well. The concentration of PI3 Kinase p110 beta can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty