Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Deoxypyridinoline ELISA Kit

Supplier: Antibodies.com LLC
A78001-96
76742-530EA 605 USD InStock
76742-530
Mouse Deoxypyridinoline ELISA Kit
Assays ELISAs
Mouse Deoxypyridinoline ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Deoxypyridinoline in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 3.125 - 200 ng/ml
  • Sensitivity: 1.875 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Mouse Deoxypyridinoline ELISA kit (A78001) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Deoxypyridinoline in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Deoxypyridinoline antigen. During the incubation, Deoxypyridinoline present in the samples or standards competes with the fixed amount of immobilized Deoxypyridinoline for binding sites on the Biotinylated Anti-Deoxypyridinoline Antibody. The more Deoxypyridinoline present in a sample or standard, the less Biotinylated Anti-Deoxypyridinoline Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Deoxypyridinoline Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Deoxypyridinoline present in each sample or standard. The concentration of Deoxypyridinoline can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty