Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat Cyp2e1 ELISA Kit

Supplier: Antibodies.com LLC
A77956-96
76743-194EA 605 USD InStock
76743-194
Rat Cyp2e1 ELISA Kit
Assays ELISAs
This Rat Cyp2e1 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat Cyp2e1 in plasma, tissue homogenates and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 3.125 to 200 ng/ml
  • Sensitivity: 1.875 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Rat Cytochrome P450 2E1 ELISA Kit (A77956) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Cytochrome P450 2E1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cytochrome P450 2E1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cytochrome P450 2E1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cytochrome P450 2E1 Antibody, which binds the captured Cytochrome P450 2E1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cytochrome P450 2E1 captured in each well. The concentration of Cytochrome P450 2E1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty