Would you like to log in again?
Porcine Acylated Ghrelin ELISA Kit
Supplier: Antibodies.com LLC
A247163-96
76743-988EA 705 USD InStock
76743-988
Porcine Acylated Ghrelin ELISA Kit
Assays ELISAs
Porcine acylated ghrelin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of porcine acylated ghrelin in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Porcine Acylated Ghrelin ELISA kit (A247163) employs the competitive enzyme immunoassay technique for the quantitative measurement of porcine Acylated Ghrelin in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Acylated Ghrelin antigen. During the incubation, Acylated Ghrelin present in the samples or standards competes with the fixed amount of immobilized Acylated Ghrelin for binding sites on the Biotinylated anti-Acylated Ghrelin antibody. The more Acylated Ghrelin present in a sample or standard, the less Biotinylated anti-Acylated Ghrelin antibody that binds to the plate. Following incubation, unbound Biotinylated anti-Acylated Ghrelin antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Acylated Ghrelin present in each sample or standard. The concentration of Acylated Ghrelin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty