Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Tyk-2 ELISA Kit

Supplier: Antibodies.com LLC
A75930-96
76746-672EA 605 USD InStock
76746-672
Human Tyk-2 ELISA Kit
Assays ELISAs
This Human Tyk-2 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Human Tyk-2 in plasma, tissue homogenates and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.313 to 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample type: Serum. plasma. tissue homogenates. and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty