Would you like to log in again?
SMART Digest™ RNase Kits
Supplier: Thermo Scientific
SMART Digest™
60120-102 60120-101
76748-630EA 1309.8 USD InStock
76748-630 76748-628
SMART Digest™ RNase Kits
Nucleic Acid Reagents RNAi
Obtain high-quality analytical results from mRNA digestions using Thermo Scientific™ SMART Digest™ RNase Kits.
  • Fast and simple mRNA digestion
  • High reproducibility
  • High sensitivity
  • High levels of data quality compatible with automation
  • A simple process and control of digestion time ensure required fragment length for comprehensive mRNA sequencing by HRAM-MS
  • Easily automate the entire protocol for high-throughput processing
  • Single nuclease digestion allows full characterization of mRNA
  • Increased reproducibility over existing protocols
  • Higher data confidence
  • No nuclease contamination of the analytical column
A significant advance in sample preparation for oligonucleotide mapping and quantitation.
These RNase kits use optimized, heat stable enzymes on magnetic beads, allowing for control of partial digestion to produce required fragments of ideal length for sequencing by HRAM-MS.
RNase kits are available with with two endoribonuclease enzyme specificities:
RNase T1 degrades single-stranded RNA at G residues
RNase A degrades single-stranded RNA at C and U residues
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty