Would you like to log in again?
Accucold® AIWD282 Water Filter Cartridge Replacement
Supplier: Felix Storch Inc
Accucold®
FILTER282REP
76748-646EA 714 USD InStock
76748-646
Accucold® AIWD282 Water Filter Cartridge Replacement
Water Dispensers
Replacement cartridge for the commercially approved two-stage water filtration system for use with the 76748-636, 76748-632, and 76748-640 (AIWD282, AIWD160 and AIWD450).
  • Commercially approved to NSF-469 standards
  • Two-stage filtration system to prevent hard water with carbon filtration to ensure fresh-tasting water
  • Easy cartridge change-out for minimal user maintenance
The FILTER282REP is a replacement cartridge for AIWD water filtration system. The cartridge-based system, allows quick replacement with minimal user maintenance.
Note: Upon installation, allow the water to run for 5 minutes prior to dispensing ice for ideal performance.
Delivery information: They are delivered with 100% guarantee.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty