Would you like to log in again?
Accucold® AIWD Water Filtration System
Supplier: Felix Storch Inc
Accucold®
FILTER282TWIN
76748-648EA 805 USD InStock
76748-648
Accucold® AIWD Water Filtration System
Water Dispensers
Commercially approved two-stage water filtration system for use with the AIWD ice/water dispensers to prevent scale formation and ensure clear, great tasting water.
  • Commercially approved to NSF-469 standards
  • Two-stage filtration system to prevent hard water with carbon filtration to ensure fresh-tasting water
  • Connects to any ³/₈" cold water line
  • Easy cartridge change-out for minimal user maintenance
The FILTER282TWIN is commercially approved to NSF-169 standards. It is a two-stage water filtration system for use with the AIWD282, AIWD160, or AIWD450 (76748-636, 76748-632 or 76748-640).
The first stage introduces an FDA approved treatment to control scale formation before entering the icemaker, which helps prevent clogging and mechanical issues. The second stage is carbon filtration that improves the taste of the drinking water, ensuring that ice and water is clear.
Note: Upon installation, allow the water to run for 5 minutes prior to dispensing ice for ideal performance.
Delivery information: They are delivered with 100% guarantee.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty