Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Environmental Test Chambers

Supplier: Avantor
VWR®
9020-0484 9020-0485 9020-0486
76777-104EA 14687.7 USD InStock
76777-104 76777-106 76777-108
VWR® Environmental Test Chambers
Testing Chambers Environmental Test Chambers
The VWR® environmental test chambers are ideal for long-term testing and accelerated stability testing of pharmaceutical products in accordance with ICH Q1A guidelines. The corrosion-free and condensation-free interior ensures exceptionally stable temperature and moisture levels, even when the chamber is fully loaded.

  • Humidity regulation with capacitive humidity sensor and corrosion-free steam humidification
  • Stainless steel interior and superior airflow design
  • LCD to display temperature and humidity along with additional information and alarms
  • Internal data logger, measured values can be read out in open format via USB
  • Access port with silicone plug, 30 mm, left
  • All models have robust twin casters with brakes

The VWR® environmental test chambers feature a stainless steel interior and steam humidification which assures reliable and corrosion-free operation at all times.

Climate homogeneneity is a given thanks to superior double-sided airflow design as well as precise temperature and humidity control. A benefit of this is the maximum usable space and greater load compared to competitor units.

The cost-effective VWR® Environmental Test Chambers offer an ethernet interface, are easy to program, make documenting test results simple and include an independent, container-based water supply.

Stability RH: ±1.5%

Certifications: UL Certified.

Delivery information: Supplied with two stainless steel racks, including shelf supports.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty