Would you like to log in again?
Live or Dead™ Yeast Viability Assay Kit
Supplier: AAT Bioquest
22475
76779-024EA 366.53 USD InStock
76779-024
Live or Dead™ Yeast Viability Assay Kit
Assays Cellular Assays
Differentiates live yeast with intact membranes will stain fluorescent green, and dead yeast with damaged membranes will stain fluorescent red.
  • robust protocol
  • live yeast will glow green
  • dead yeast will glow red
This kit contains two ready-to-use nucleic acid stains: MycoLight™ Green JJ98, a replacement for SYTO™ 9, and propidium iodide.
Quantitatively distinguishes live and dead yeast within minutes using flow cytometry
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty