Would you like to log in again?
ReadiLink™ DIG (Digoxigenin) Oligo and ssDNA Labeling Kit
Supplier: AAT Bioquest
ReadiLink™
17492 17493
76779-050EA 374.7 USD InStock
76779-050 76779-064
ReadiLink™ DIG (Digoxigenin) Oligo and ssDNA Labeling Kit
Protein Linking and Labeling Systems
The kit is optimized for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
  • Efficient labelling
  • Contains all essential reagents
  • Multiple applications
The resulting DIG-labeled oligo and ssDNA probes are ideally suited for a variety of biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Packaging: Packaged in plastic vials.
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty