Would you like to log in again?
ReadiUse™ 6-Color Human TBNK Antibody Kit
Supplier: AAT Bioquest
90011 90010
76779-068EA 1007.11 USD InStock
76779-068 76779-112
ReadiUse™ 6-Color Human TBNK Antibody Kit
Assays Cellular Assays
Each TBNK dried reagent contains a cocktail of fluorescently-labeled monoclonal antibodies directed against human CD3, CD4, CD8, CD16, C19, CD45, and CD56.
  • dry reagent format
  • no wash
  • easy sample preparation
The kit is designed to be used in a “lyse/no-wash” format for quick and easy sample preparation, with less inconsistencies.
Also included in this kit is a viability stain to identify dead cells and a precise number of fluorescent counting beads to determine absolute lymphocyte subset cell concentrations.
Kit for flow cytometry utilizes dried multicolor beads to delineate principal lymphocyte subsets - CD4+ T cells, CD8+ T cells, B cells, and NK cells – in whole blood specimens
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty