Would you like to log in again?
ReadiLink™ Cy5 Nick Translation dsDNA Labeling Kit
Supplier: AAT Bioquest
ReadiLink™
17408 17409
76779-072EA 406.32 USD InStock
76779-072 76779-042
ReadiLink™ Cy5 Nick Translation dsDNA Labeling Kit
Protein Linking and Labeling Systems
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearized plasmid DNA.
  • Provides all necessary reagents
  • Adjustable ratios
  • Wide sample type compatibility
The resulted Cy5-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridization (FISH).
Packaging: Packaged in plastic vials.
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty