Would you like to log in again?
Live or Dead™ Yeast CFDA-AM/Propidium Iodide Vitality Kit
Supplier: AAT Bioquest
22476
76779-096EA 366.53 USD InStock
76779-096
Live or Dead™ Yeast CFDA-AM/Propidium Iodide Vitality Kit
Assays Cellular Assays
Differentiates live yeast with intact membranes will stain fluorescent green, and dead yeast with damaged membranes will stain fluorescent red.
  • robust protocol
  • live yeast will glow green
  • dead yeast will glow red
The kit uses CFDA-AM and Propidium Iodide. CFDA-AM is a cell permeable esterase substrate.
The acetoxymethyl (AM) group of the substrate will allow CFDA to penetrate cell membranes. Once inside the cells, lipophilic blocking and diacetate groups are cleaved by esterases, resulting in green fluorescence. In contrast, Propidium Iodide only stain the yeasts with damaged membranes resulting in red fluorescence.
Convenient way to test viability of yeasts
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty