Would you like to log in again?
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Supplier: AAT Bioquest
Bucculite™
22328
76779-110EA 537.56 USD InStock
76779-110
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Assays Cellular Assays
Provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay
  • alternative to anti-BrdU
  • extremely sensitive
  • deep red fluorescence
The resulted iFluor® 647-labeled DNA formed in cells is imaged with a Cy5 filter set.
Uses XdU which is incorporated into cellular DNA during DNA synthesis
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty