Would you like to log in again?
TableWorx Performance Work Tables
Supplier: Metro International
TableWorx
TWM2430SU304BK TWS3072SU316BS TWM3072SI304K TWS3072FS316BS TWM2448SU304BK TWM3636SU304BK TWM2472SU304K TWM3036SI304K TWM3096FS304BK TWS3636SU316S TWS2496SU304BK TWM3660SI316S TWS3636SI304BS TWS2460SU304S TWM3072SU304BS TWM2424FS304BK TWS2448SU304K TWM3660SU304BS TWM3072SI304S TWS2496SI304BK TWS3048SU304K TWS3036SI316S TWM3060SI316BS TWS2496FS304BK TWS2496FS304K TWM3072FS304K TWS3648FS304K TWS2496FS316S TWS2424SU304S
76779-142EA 1662.11 USD InStock
76779-142 76779-980 76779-782 76780-234 76780-156 76780-254 76779-634 76779-978 76779-554 76779-972 76779-556 76779-274 76779-574 76779-992 76779-350 76780-104 76779-708 76779-704 76779-968 76779-926 76779-888 76779-202 76780-062 76780-260 76779-564 76779-542 76779-446 76780-180 76779-522
TableWorx Performance Work Tables
Furniture Tables and Workbenches
Metro TableWorx is a truly modular platform of performance work tables designed to allow you to customize your perfect solution.
  • Innumerable options and a truly modular platform allow you to customize a perfect solution
  • Overhead riser solution can be added at anytime converting it into a productivity workcenter
  • Work surface alloy: type 304 or 316 stainless steel
  • Work surface load capacity of 650 lbs.
  • Height adjustability
All TableWorx Work Tables feature a 16-gauge, Type 304 or 316 stainless steel work surface reinforced with 16-gauge hat channels. The sound deadened work surface features 90-degree turned down edges on all four sides.
Out-of-the box, TableWorx can be assembled to any one of six working surface heights: 34" (864 mm), 35" (889 mm), 36" (914 mm), 37" (940 mm), 38" (965 mm), and 39" (991 mm). Stationary work table legs include stainless steel leveling bullet feet with 1.125" (29 mm) range of adjustability. Mobile-ready tables are delivered without casters.
Elevate your efficiency to new levels with the addition of TableWorx productivity riser. Risers can be easily added to any TableWorx work table. A full array of overshelves, wire grids, and accessories can be added to maximize every inch of space, making you more comfortable and productive than ever before. Risers can be mounted at the rear or center of the work table.
Certifications: NSF Certified.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty