Would you like to log in again?
VWR® Full Size Sliding Wire Basket for Select Refrigerator Units
Supplier: VWR
VWR®
VWR-BSKT-BASE-FIXED-0504 VWR-SLD-BSKT-FULL-0504 VWR-BSKT-BASE-FIXED-10.5 VWR-SLD-BSKT-FULL-10.5
76780-330EA 133.1 USD InStock
76780-330 76780-334 76780-332 76780-336
VWR® Full Size Sliding Wire Basket for Select Refrigerator Units
Baskets Wire Baskets
VWR® sliding storage baskets can streamline inventory management and enhance the safety and security of your products improving overall efficiency and productivity.
  • Enhanced organization
  • Improved air circulation
  • Maximized storage space
Baskets are designed to fit 10.5 cu. ft. compact refrigerators and 5.2 cu. ft. freestanding undercounter refrigerators. Their sturdy construction allows you to stack and organize inventory for efficiency and easy accessibility.
Baskets are designed to promote superior air circulation, preventing hot spots and maintaining consistent temperatures throughout the chamber. This improves the shelf life of your products and reduces the risk of spoilage.
The bottom mounted fixed basket can add extra usable space to maximize your unit storage.
Accessories information: These VWR® Full size sliding wire baskets are compatible accessories for selected VWR® refrigerator units.
Ordering information: Full size baskets are factory installed only - See disclaimer.
Caution: Disclaimer: Full sized baskets are a factory-installed accessory. Bottom mounted baskets can be manually installed. The unit and the basket(s) must be ordered together. 10.5 cu. ft. units can replace up to 6 full sliding Baskets and 1 half-size, bottom-mounted fixed basket. 5.2 cu. ft. freestanding undercounter units can replace up to 3 full sliding Baskets and 1 half-size, bottom-mounted fixed basket. Lesser basket quantities can be ordered for each refrigerator and will be installed from the bottom of the unit upwards and equally spaced.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty