Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylammonium iodide 97%

Supplier: Ambeed
Warning
Synonyms: TBAITetra-n-butylammonium iodide

A169033-100G A169033-1000G A169033-25G A169033-5G A169033-500G
76784-146EA 128.75 USD InStock
76784-146 76784-148 76784-150 76784-154 76784-152
Tetrabutylammonium iodide 97%
Tetrabutylammonium iodide
Caution: for Research Use Only


Formula: C₁₆H₃₆IN
MW: 369.37 g/mol
MDL Number: MFCD00011636
CAS Number: 311-28-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty