Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(2-ethylhexyl)phosphate 98%

Supplier: Ambeed
Warning
Synonyms: Trioctyl phosphateTris(2-ethylhexyl) phosphatePhosphoric acid tris(2-ethylhexyl ester)

A411328-100G A411328-500G A411328-25G A411328-1000G
76789-026EA 35.02 USD InStock
76789-026 76789-020 76789-022 76789-024
Tris(2-ethylhexyl)phosphate 98%
Tris(2-ethylhexyl) phosphate
Caution: for Research Use Only


Formula: C₂₄H₅₁O₄P
MW: 434.64 g/mol
MDL Number: MFCD00009491
CAS Number: 78-42-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty