Would you like to log in again?
Sterilization Wraps, 2-Ply SMS, One-Step Heavy Duty
Supplier: Keystone Cleanroom Products
KCRP1-300-5454 KCRP1-300-5472 KCRP1-300-1818 KCRP1-300-0808 KCRP1-300-2424 KCRP1-300-4548 KCRP1-300-1212 KCRP1-300-3636 KCRP1-300-5490 KCRP1-300-4545 KCRP1-300-0912 KCRP1-300-4040 KCRP1-300-0909 KCRP1-300-4848 KCRP1-300-3030 KCRP1-300-6080 KCRP1-300-6060 KCRP1-300-2020 KCRP1-300-1515
76795-478CS 734.4 USD InStock
76795-478 76795-468 76795-476 76795-498 76795-466 76795-488 76795-474 76795-496 76795-486 76795-464 76795-472 76795-494 76795-484 76795-462 76795-470 76795-492 76795-482 76795-490 76795-480
Sterilization Wraps, 2-Ply SMS, One-Step Heavy Duty
Sterilization Wraps
One-Step super duty SMS 2-ply wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
  • Manufactured and packaged in a certified ISO 8 Cleanroom
  • 2-ply
  • Clean - no cellulose
  • Strong, rip and tear resistant 88 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilization gases, but prevents liquid and microbial penetration
  • Gram weight: 50 (each layer)
For use in steam, ethylene oxide and hydrogen peroxide sterilization processes.
Delivery information: Certificate of Conformance sent with each shipment.
Packaging: Sub packed into sealed labeled poly bags with lot number and expiration date for complete traceability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty