Would you like to log in again?
Hot Plate Stirrers, AREX 5
Supplier: VELP Scientifica
SA20510570 F20510570
76795-554EA 645.81 USD InStock
76795-554 76795-560
Hot Plate Stirrers, AREX 5
Stirrers Magnetic Hotplate Stirrers
The AREX 5 is a robust and versatile hotplate stirrer suitable for applications ranging from basic to temperature-sensitive where precise sample thermoregulation is required.
  • Easy-to-use and safe
  • Ensures a high level of safety and protection for laboratory operators
  • Two illuminated indicators always inform the operator when the heating is on
  • The 135 mm diameter top is protected from chemicals and scratches by the superior resistant CerAlTop™ ceramic coating
The AREX 5 can work stand-alone or be upgraded to digital by connecting the VTF digital temperature controller. The CerAlTopTM plate reaches a maximum temperature of 310 °C; when connected with the VTF, the AREX 5 is able to thermoregulate the medium up to 300 °C with an accuracy of ±0.5 °C.
The internal electronic parts are protected by the insulating disk, which, together with the robust die-cast aluminum housing, enables the AREX 5 to withstand even the most demanding laboratory environments and applications, such as when working with cold media (below 0 °C).
Certifications: IP42 certified.
Delivery information: It is backed up by a 3-year warranty (online product registration required).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty