Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digital Hot Plate Stirrers, AREX 5

Supplier: VELP Scientifica
SB20510580 F20510580 SC20510580 SA20510580
76795-566EA 880.65 USD InStock
76795-566 76795-556 76795-564 76795-562
Digital Hot Plate Stirrers, AREX 5
Stirrers Magnetic Hotplate Stirrers
The AREX 5 Digital is the perfect solution for temperature-sensitive laboratory applications where precise and accurate thermoregulation of the sample is required to ensure maximum reproducibility.

  • A safe hotplate stirrer designed to last
  • Ensures a high level of safety and protection for laboratory operators
  • Internal electronic parts are protected by the insulating disk
  • 135 mm diameter top is protected from chemicals

The CerAlTopTM plate reaches a maximum temperature of 310 °C; when connected with the temperature probe (Pt100 and Pt1000) the AREX 5 Digital is able to heat up the medium up to 310 °C with an accuracy of ±1.0 °C while with the VTF the AREX 5 Digital is able to heat up the medium up to 300 °C with an accuracy of ± 0.5 °C.

When the hotplate stirrer is connected to a temperature probe, it is possible to choose 2 different thermoregulation modes to match all requirements:
FAST: the Hot Plate Stirrer quickly reaches the set point temperature with a little overshoot
FINE: slower temperature set point reaching with an optimized overshoot

The AREX 5 Digital is equipped with VELP-engineered SpeedServo™ torque compensation technology that ensures constant speed even when the viscosity of the medium changes. The strong magnetic coupling supports reactions with multiple vials ensuring constant speed in all positions, visit our accessories section for more details.

Certifications: IP 42 certified.

Delivery information: It is guaranteed by a 3-year warranty (online product registration required).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty