Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Flexible 6 Routine Clinical Centrifuge

Supplier: VWR
VWR®
00-376-037-000
76795-578EA 1181.46 USD InStock
76795-578
VWR® Flexible 6 Routine Clinical Centrifuge
Centrifuges Benchtop Centrifuges
Completely customizable to suit the unique needs of any routine laboratory. Monitor remaining cycle time on the digital display and program cycle parameters on the fly.

  • View time remaining and set cycle parameters on the fly using the digital display
  • Use default presets for chemistry, coag, and urine; or store up to 10 custom time, speed, acceleration, and braking settings
  • LED lid lighting system indicates whether the centrifuge is ready (off), running (on), or done (flashing) and continues flashing until the lid is opened, preventing forgotten tubes in even the noisiest labs
  • Max speed 2000×g/3800 rpm
  • Spin up to 6 tubes (75 to 100 mm/3 to 10 ml) out of the box, no additional accessories needed
  • 2 year warranty (lifetime on rotor)
  • Lightweight and compact, the Flexible 6 is ideal for smaller labs and can be moved around to address bottlenecks

Customize to suit the unique needs of any routine laboratory with the VWR® Flexible 6 Clinical Centrifuge. Monitor cycle time remaining on the digital display and program cycle parameters (runtime, rpm, g-force, and more) on the fly. The LED lid lighting system indicates whether the centrifuge is ready (off), running (on), or done (flashing) and continues flashing until the lid is opened. Lightweight and portable, the VWR® Flexible 6 Clinical Centrifuge is ideal for smaller laboratories and can be moved around to address bottlenecks. Max speed 2000×g/3800 rpm.

Certifications: CE, MET, UKCA.

Delivery information: Supplied with horizontal rotor and universal tube holders.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty