Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® STAT Coag Clinical Centrifuge for STAT Coag Processing

Supplier: VWR
VWR®
00-676-037-000
76795-588EA 4036.98 USD InStock
76795-588
VWR® STAT Coag Clinical Centrifuge for STAT Coag Processing
Centrifuges Benchtop Centrifuges
Compact benchtop STAT centrifuge with fixed-angle rotor for rapid COAG/PPP processing. Simple to operate with status tracker lid lighting.

  • 3 convenient presets for common STAT cycles, select using simple two-button interface
  • LED lid lighting system indicates cycle status: Off when ready, on when running, flashing when done
  • Max speed 6600 rpm/4400×g
  • 12 tube capacity (75 mm/4 ml)
  • Compact size to be used in an array in accessioning, further driving down TAT
  • Fixed angle rotor design for optimal sample processing
  • 2 year warranty

Optimize for PPP with the VWR® STAT Coag Clinical Centrifuge. Designed with platelet poor plasma spins in mind, the VWR® STAT Coag offers three convenient presets for common laboratory needs. Its fixed angle rotor spins up to 12 teal-top 75 mm/4 ml tubes at a time. With a compact footprint and simple two-button controls, the VWR® STAT Coag is right at home in accessioning, or for tube processing in the emergency department. Max speed 4400×g/6600 rpm.

Certifications: CE, MET, UKCA.

Delivery information: Supplied with fixed angle rotor.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty