Would you like to log in again?
Parasep® Gold Standard Fecal Parasite Concentration, Apacor
Supplier: Apacor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
108100 108800 149910 149001 145000 149970 108900 146001 108801 146300 249300 149920 145300 108810 108920 108000 160001 149995 149901 149000 249400 249420 146000 149900
10048-638BX 47.2 USD InStock
10048-638 10048-604 10048-714 10048-718 76066-146 10183-880 10048-716 10770-304 10183-870 10184-190 10860-668 76795-688 10048-670 76795-686 10048-690 76795-684 10796-704 76795-690 76498-754 10796-702 10048-678 76498-752 10048-676 10048-664
Parasep® Gold Standard Fecal Parasite Concentration, Apacor
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection
Parasep fecal parasite concentrators are for fecal concentration of helminth ova and larvae/protozoa cysts and oocysts.
Parasep's patented vertical filter device removes blockage, which occurs in all flat filter methods, thus increasing the yield of parasites. Sample clarity is enhanced too with our dual 425 µm and 220 µm filtration methodology, as it reduces the amount of debris on the slides making them easier to examine.
We have many users of our Parasep methodology in the US, including the large parasitology reference laboratories ARUP and Mayo Clinic. Apacor is proud to confirm that our collaborators in the US are ARUP with whom we have improved our reagents and procedure further still. On their web site you will be able to review some of the benefits of our procedure, a training video and their IFU for Parasep.
Current techniques to identify intestinal parasites in patients can be a laborious and time-consuming process. Parasep eliminates the cause of these issues improving processing time by 174% against the closest competition and saving 80% time during the centrifugation process.
With the human resources optimized by switching to Parasep, Biomedical scientists can utilize their time working on other areas in the lab. The increased sensitivity of the test will improve patient care and the speed in which the test can be run will enable Biomedical scientists to use their time on other tests. Request a sample today and see the benefits for yourself!
Certifications: FDA and CE certified.
Ordering information: Parasep is available in a variety of pre-dispensed combinations. Our unique, patented, vertical filtration, parasite fecal concentration device is also available without reagent.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty