Would you like to log in again?
Optimum Growth™ Inversion Transfer Caps
Supplier: Thomson Instrument
931710-4 931594-4 931704-8 931618-8 931705-4 931708-4 931706-4 931598-4 931595-4 931598-4 931708-4 931595-4 931616-4 931614-8 931704-8 931706-4 931705-4 931596-4 931618-8 931470-8 931596-4 931702-8 931616-4 931702-8 931614-8 931594-4 931710-4
76808-340CS 537.33 USD InStock
76808-340 100035-978 100035-974 76808-322 76808-344 76808-342 76808-320 76808-348 76808-346 76808-324 76808-328 102963-032 102962-984 102962-980 102830-244 102963-024 100035-986 76808-332 76808-336 100035-982 76808-334 102963-020 76808-338 102962-976 102962-978 77552-892 102962-974
Optimum Growth™ Inversion Transfer Caps
Closures Safety Caps and Safety Waste Caps
The sterile optimum Growth™ transfer caps and inversion transfer caps allow for a simple aseptic transfer of media or cells to cell bags, bioreactors, and flasks of all makes.
These caps are designed for use with the Optimum Growth™ flask product line. Simply replace the culture cap with the transfer cap and connect to the vessel of choice.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty