Would you like to log in again?
Optimum Growth™ Flask
Supplier: Thomson Instrument
Optimum Growth™
931116 931116 931114-PORT-TRT 931117 931113 931111 931111 931114 931110 931112 931110 931113 931114 931112 931110-11
102908-366CS 659.4 USD InStock
102908-366 76808-394 76808-392 76808-398 76808-396 76808-400 102830-248 77552-880 77552-882 102830-246 102830-250 77552-884 76808-390 101641-616 103244-092
Optimum Growth™ Flask
Flasks Cell Culture Flasks
The Optimum Growth™ flasks provide excellent growth with space saving capability.
  • Baffles designed for high aeration and low shear
  • Same footprint as comparable Fernbach flask
  • Less foaming than disposable Fernbach
  • Transfer cap option connects directly to cell bags or bioreactors with quick connect, luer lock or tube fusing
  • 0.2 μm vented cap
  • Individually packaged and sterilized
They are designed in a way that protein production will scale consistently across all sizes, unlike any other shake flasks on the market. Additionally, these flask features allow for consistent shake speeds from the 125 ml up to the 7 L flasks.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty