Would you like to log in again?
Ultra Yield™ Flasks, Thomson Instrument Company
Supplier: Thomson Instrument
931136-B 931138 931138 931141 931141 931147 931147 931144 931144 931136-B
76808-408CS 504.26 USD InStock
76808-408 76808-410 101431-642 101431-654 76808-402 101431-644 101431-646 76808-406 103537-308 76808-404
Ultra Yield™ Flasks, Thomson Instrument Company
Flasks Cell Culture Flasks
Thomson's Ultra Yield™ Flasks are so named because these flasks actually enhance the growth of E.coli as well as other microbial cells.
The AirOtop® Seals are designed as a single-use, disposable seal. Enhanced AirOtop® Seals offer simple, disposable, sterile high air exchange. Seals fit all shake flasks. The Enhanced AirOtop® Seal's properties include a 0.2 um sterile barrier with a long-term resealable membrane for use with Ultra Yield™ and other brand flasks. Testing has been conducted on multiple customer samples with great results on up to 7 days of cellular growth. Testing included Protista (Algae), E.coli, and other microbes resulting in improved growth.
The patented design of the flasks renders them to an improved microbial cell yield. Flasks are available in standardized sizes of 125 ml, 250 ml, 500 ml, and 2.5 l. The flasks can be closed snug using the AirOTop seals. These seals are designed to fit on the flask-tops. Aeration caps are also available for long-term use on Flasks for applications such as CHO and other suspension cell growth procedures.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty