Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Standard|Filter Vials, Thomson Instrument Company

Supplier: Thomson Instrument
35541-200 35539-200 35541-500 35540-200 35540-500 35541-500 35535-200 35531-500 35540-200 35531-200 35531-200 35538-200 35530-200 35539-500 35539-500 35530-500 35540-500 35539-200 35541-200 35538-500 35530-500 35531-500 35538-200 35535-200 35530-200 35535-500 35535-500 35538-500
101623-354CS 649 USD InStock
101623-354 76808-622 76808-620 101623-336 76808-626 101623-358 101623-334 101623-356 76808-624 101447-036 101623-338 76808-628 101447-038 101623-340 76808-630 101623-360 76808-634 76808-612 101623-344 101623-342 76808-610 76808-632 76808-616 101623-348 76808-636 101623-346 76808-614 76808-618
Standard|Filter Vials, Thomson Instrument Company
Filters Syringeless Filters
Recommended for samples containing less than 10% solid particulates.

  • Simply add sample, diluent, and press
  • Color-coded caps allow easy identification of 0.2 or 0.45 μm membranes in PVDF, PTFE, PES, or Nylon
  • Fits most standard 12×32 mm autosamplers

Standard|Filter Vials are designed to speed up sample prep and minimize transfer steps. Available in a variety of membranes and pore sizes. Simply add sample and diluent, press, and it is ready for analysis. This process minimizes sample loss. Pre-slit caps ensure a no-hassle, clean withdraw.

The Standard|Filter Vials are made from polypropylene and come in a variety of membranes and pore sizes.

Accessories information: The accessory Filter Vial Presses quickly depress autosampler-ready filter vials.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty