Would you like to log in again?
BD MAX™ ExK™ DNA 3 (Urine/Swab (UTM))
Supplier: BD Diagnostics
BD MAX™
442821
76808-684EA 1131.56 USD InStock
76808-684
BD MAX™ ExK™ DNA 3 (Urine/Swab (UTM))
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
BD MAX™ ExK™ DNA 3 (Urine/Swab (UTM)) is for open system reagents to be used on the BD MAX™ System. DNA extraction from urine and UTM swab.
  • DNA extraction from urine and UTM swab specimens
  • Used for open system capabilities
  • Use with the BD MAX™ System
BD offers an array of nucleic acid extraction and amplification reagents for use on the BD MAX™ System. Worldwide BD MAX™ System assay development partners and end-users are able to create User-Defined Protocols (UDPs) by leveraging these reagents to meet emerging diagnostic demands and address regional healthcare needs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty