Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MultiPlate Genie Microplate Shaker, 230 V

Supplier: Scientific Industries
MultiPlate Genie
SI-4101
76808-696EA 1015.82 USD InStock
76808-696
MultiPlate Genie Microplate Shaker, 230 V
Shakers and Mixers Microplate Shakers
Fully programmable microplate shaker for four plates (8 with optional tray).

  • 230 V model
  • Provides consistent well-to-well, plate-to-plate mixing for 96 and 384-well microplates, deep well plates, or PCR plates
  • Accepts PCR work up racks with standard microplate footprint for use with PCR tubes and strips
  • Featuring programmable speed, time, pulsing/interval mixing - for accurate and repeatable results

Digital true vortex mixing microplate shaker handles up to 4 microplates, (or up to 8 plates with the optional accessory tray). The combined speed and 1 mm orbit provide consistant well to well and plate to plate uniformity, even in 384-well plates. Featuring programable speed, time and continuous or pulse time, it's ideal for achieving accurate and repeatable results. The Pulse feature allows rapid pulsing or timed 'on/off' mixing for interval mixing. Ideal for use in PCR preparation, digestions, bead mixing, ELISA, EIA, and colorimetric assay applications.

Pulse (Interval): 0.1 s to 99 min

Certifications: CE marked.

Delivery information: Supplied with tray for 4 microplates. An optional stacking tray accessory for additional 4 microplates (total of 8) is available to purchase separately.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (76808-694) is also available, please see separate product page.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty