Would you like to log in again?
DURAN® Bottle Carrying Systems
Supplier: DWK Life Sciences
DURAN®
292816305 292807309 292826306 292836307 292809104 292808605 292806304
76808-780CS 37.55 USD InStock
76808-780 76808-790 76808-784 76808-794 76808-782 76808-792 76808-788
DURAN® Bottle Carrying Systems
Carriers Bottle Carriers/Containers
The ergonomic design offers health and safety benefits and allows easy and safe transport of bottles.
  • Increase comfort and safety when pouring liquids from larger bottles
  • Collar is only removable by cutting, making the attachment 100% secure in use
  • Carrier can be cleaned attached to the bottle in an automated washer
  • Autoclavable
The Bottle Carrying System range fits standard laboratory bottles that have a GL 45 neck finish, and are available for 2 or 5 L bottle sizes. The carriers are ergonomically designed and offer health and safety benefits to the user. They allow the easy and safe transport of bottles around the laboratory or production facility. The robust pouring side handle on the 5 liter version makes pouring liquids from the larger bottles more controllable and safer. All materials are fully autoclavable. The collar that fits to the neck of the bottle is only removable by cutting, so the attachment is 100% secure in use.
Ordering information: Bottle is not included and must be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty