Would you like to log in again?
VWR® Thermal-Transfer Cryogenic Labels, Zebra-Compatible
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
VWR-TTC-1-6 VWR-TTC-3-5 VWR-TTC-3-8 VWR-TTC-1-3 VWR-TTC-1-5 VWR-TTC-3-7 VWR-TTC-3-2 VWR-TTC-1-7 VWR-TTC-1-1 VWR-TTC-3-4 VWR-TTC-3-6 VWR-TTC-3-3 VWR-TTC-3-1 VWR-TTC-1-4 VWR-TTC-1-2
76808-960RL 25.85 USD InStock
76808-960 76808-870 76808-864 76808-842 76808-952 76808-852 76808-962 76808-972 76808-840 76808-912 76808-878 76808-966 76808-900 76808-828 76808-948
VWR® Thermal-Transfer Cryogenic Labels, Zebra-Compatible
Labels
Thermal Transfer Cryogenic Labels are ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications.
  • Can be placed in most incubators
  • Labels are thermal transfer top coated and require a ribbon for printing
Labels feature a unique adhesive that can withstand extreme temperatures from –196 to 121 °C (–321 to 250 °F), without the need for lamination.
Ordering information: Rolls of labels come on a 1 or 3" core, depending on the intended printer model.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty