Would you like to log in again?
VWR® Cryogenic Dot and Strip Labels
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
VWR-DTC56 VWR-DTC13 VWR-DTC55 VWR-DTC25 VWR-DTC48 VWR-DTC10 VWR-DTC5 VWR-DTC46 VWR-DTC30 VWR-DTC2 VWR-DTC3 VWR-DTC6 VWR-DTC28 VWR-DTC36 VWR-DTC58 VWR-DTC19 VWR-DTC39 VWR-DTC26 VWR-DTC51 VWR-DTC9 VWR-DTC12 VWR-DTC43 VWR-DTC22 VWR-DTC33 VWR-DTC17 VWR-DTC45 VWR-DTC1 VWR-DTC57 VWR-DTC32 VWR-DTC37 VWR-DTC15 VWR-DTC49 VWR-DTC42 VWR-DTC31 VWR-DTC14 VWR-DTC24 VWR-DTC35 VWR-DTC54 VWR-DTC29 VWR-DTC23 VWR-DTC53 VWR-DTC47 VWR-DTC7 VWR-DTC34 VWR-DTC41 VWR-DTC27 VWR-DTC21 VWR-DTC52 VWR-DTC38 VWR-DTC16 VWR-DTC4 VWR-DTC11 VWR-DTC44 VWR-DTC20 VWR-DTC50 VWR-DTC18 VWR-DTC8 VWR-DTC40
76808-860RL 33.86 USD InStock
76808-860 76808-980 76808-942 76808-940 76808-862 76808-902 76808-946 76808-868 76808-944 76808-866 76808-906 76808-904 76808-826 76808-908 76808-890 76808-894 76808-970 76808-892 76808-898 76808-854 76808-974 76808-930 76808-896 76808-858 76808-934 76808-978 76808-856 76808-976 76808-932 76808-938 76808-936 76808-882 76808-880 76808-964 76808-886 76808-884 76808-968 76808-924 76808-846 76808-922 76808-888 76808-844 76808-928 76808-926 76808-848 76808-872 76808-832 76808-876 76808-874 76808-950 76808-836 76808-956 76808-834 76808-954 76808-910 76808-916 76808-838 76808-914
VWR® Cryogenic Dot and Strip Labels
Labels
VWR® Cryogenic dot and strip labels are ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications.
  • Can be placed in most incubators
  • DYMO or Zebra compatible
  • Direct thermal and cryogenic
Labels feature a unique adhesive that can withstand extreme temperatures from –196 to 121 °C (–321 to 250 °F), without the need for lamination.
Ordering information: Rolls of labels are supplied on a 44 mm Ø (1.75") core. Laser printer label sheets are 21.6W×27.9L cm (8.5×11"). Assorted labels come in blue, green, purple, red, and yellow. Roll of assorted labels contains 1000 labels of each color. Boxes of assorted laser printer labels contain five sheets of each color.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty