Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sanitary Cubelet Ice Machine and Water Dispensers

Supplier: Hoshizaki
DCM-300BAH-OS DCM-300BAH
76808-990EA 14498.18 USD InStock
76808-990 76808-988
Sanitary Cubelet Ice Machine and Water Dispensers
Ice Makers
The Hoshizaki sanitary cubelet ice machine and water dispensers feature either LED touch buttons (76808-988) or Opti-Serve touchless operation (76808-990). Both models feature a durable stainless steel exterior, advanced CleanCycle24™ design, and stainless steel auger with greaseless bearing.

  • Up to 321 lbs. of ice production per 24 hours
  • Up to 40 lbs. of built-in storage capacity (1.3 ft.³)
  • 2 second flush cycle every hour
  • Refrigerant: R-404A refrigerant
  • Power: 10.25 A, 6.84 kW
  • Electrical: 115 V, 60 Hz, 1 phase
  • Dispense ice and water

The units can produce up to 321 lbs. of easy to chew 'cubelet' ice per day and hold 40 lbs. of ice in their storage bin. Units feature a durable and easy-to-clean stainless steel exterior with H-Guard plus antimicrobial agent for an additional layer of protection. The CleanCyle automatically removes impurities from incoming water so you can ensure your customers are receiving sanitary water and ice. The units uses R-404A refrigerant and require a 115 V electrical connection for operation. Units are hardwired.

Self-contained design reduces opportunities for cross contamination. Corrosion resistant stainless steel exterior.

Certifications: NSF, UL certified, AHRI certified, ADA compliant.

Packaging: Ice machines are on a skid, packaging on all four corners, and strapped down, wrapped in plastic.

Caution: * Capacity based on 100% of total volume ×30 lbs./ft.³ average density of ice.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty