Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tissue Cassettes, Round Hole, Without Lid

Supplier: Four E's USA
K2106042
76809-040CS 671.17 USD InStock
76809-040
Tissue Cassettes, Round Hole, Without Lid
Embedding Cassettes Tissue Processing Cassettes
Round hole processing cassettes ensure precise tissue processing with minimized deformation.

  • For use with reusable metal stainless steel lids (sold separately)
  • 45° angled writing surface for convenient labeling
  • For use with laser printers

Crafted for optimal fluid exchange during processing, these round hole cassettes maximize efficiency in tissue preparation.

Packaging: Bulk packaging: 500 per bag, 10 bags per case.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty