Would you like to log in again?
Thermo Scientific™ SoftFit-L™ Pipette Tips
Supplier: Thermo Fisher Scientific
Thermo Scientific™ SoftFit-L™
2769RC-HR 2789RC-HR 3722RC-HR 3752RC-HR 2779RC-HR 3751RC-HR 3782RC-HR 2739RC-HR 3742RC-HR 3781RC-HR 3731RC-HR 3732RC-HR 3721RC-HR 2749RC-HR
76809-090CS 982.87 USD InStock
76809-090 76809-092 76809-094 76809-086 76809-130 76809-132 76809-088 76809-120 76809-122 76809-124 76809-116 76809-126 76809-118 76809-128
Thermo Scientific™ SoftFit-L™ Pipette Tips
Pipette Tips
Thermo Scientific SoftFit-L™ pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.
  • Recycle nonhazardous pipette tips, racks, and associated packaging with The Tipcycle Program. Sustainable design based on virgin resins
  • Guaranteed free of detectable DNA, DNase/RNase, endotoxins and heavy metals
  • Tested for endotoxin (pyrogen) contamination
  • Resins are pre-tested for metal contamination
  • Graduations allow a visual check of pipetting accuracy
Perform pipetting accurately and consistently with Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips, which seal when exposed to potential contaminants. The pipette tips from the reload inserts clip securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in Thermo Scientific™ MicroPoint™, extended length, low retention.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty