Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylcyclohexanone 97%

Supplier: Ambeed
Danger

A596950-5G A596950-25G
76819-366EA 5.15 USD InStock
76819-366 76819-364
4-Methylcyclohexanone 97%
4-Methylcyclohexanone
Caution: for Research Use Only


Formula: C₇H₁₂O
MW: 112.17 g/mol
MDL Number: MFCD00001643
CAS Number: 589-92-4
UN: 1224
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty